Order Entry
Canada
Orders LinkContactUsLinkComponent
Cuvette Stirring Rods, FireflySci

Cuvette Stirring Rods, FireflySci

Cuvette stirring rod for semi-micro cuvettes
Catalog # 76016-686
Supplier:   FIREFLYSCI, INC.
Supplier #:  P68
Cuvette Stirring Rods, FireflySci
Catalog # 76016-686
Supplier:  FIREFLYSCI, INC.
Cuvette Stirring Rods, FireflySci
Cuvette Stirring Rods, FireflySci

Cuvette stirring rod for semi-micro cuvettes

Price
$243.36
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Stirring rods for the manual stirring of samples in a 10 mm cuvette.

  • Steam autoclavable at 121 °C (250 °F)

Stirring rods allow samples to be continuously spun at the bottom of a cuvette to prevent sedimentation and coagulation.

Specifications
  • Description:
    Cuvette stirring rod for semi-micro cuvettes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.