Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Hardshell Plastic Cuvette Cases, Type P

Cuvette case for 45 - 55 mm cells, Plastic
Catalog # 76016-652
Supplier:   FIREFLYSCI, INC.
 :  CASEP4
Hardshell Plastic Cuvette Cases, Type P
Catalog # 76016-652
Supplier:  FIREFLYSCI, INC.
Hardshell Plastic Cuvette Cases, Type P
Hardshell Plastic Cuvette Cases, Type P

Cuvette case for 45 - 55 mm cells, Plastic

Price
$75.09
Log in for availability
Each
Product Details & Documents

Cuvette cases hold 4 cells up to 10 mm L/P each, or two 20 mm L/P cells, or one 50 mm L/P cell.

  • Black opaque finish
Specifications
  • Description:
    Cuvette case for 45 - 55 mm cells
  • Material:
    Plastic