Order Entry
Canada
Orders LinkContactUsLinkComponent
Shimadzu Type 16 Multi-Micro Cell Array, FireflySci

Shimadzu Type 16 Multi-Micro Cell Array, FireflySci

UV Quartz, 50 µl (16x), 5 mm
Catalog # 76015-214
Marketsource
Supplier:   FIREFLYSCI, INC.
Supplier #:  16UV5
Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
Catalog # 76015-214
Supplier:  FIREFLYSCI, INC.
Marketsource
Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
Shimadzu Type 16 Multi-Micro Cell Array, FireflySci

UV Quartz, 50 µl (16x), 5 mm

Price
$3,021.36
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Multi-Micro Cell Array has 16-chambered multi-micro array of cells for doing multiple measurements.

  • Highly accurate results, only need to test sample once
  • Made by hand, ensuring high quality optical polishing with no defects
  • Fire-fused to ensure that cuvettes are robust and durable
  • Comes in UV Quartz
  • Available in 10 mm lightpath
  • Two windows optically polished
  • Walls are made from black quartz to block out stray light

Typically used in Shimadzu machines.

Each cell goes through a rigorous inspection process before leaving the factory.

Specifications
  • Volume:
    50 µl (16x)
  • Description:
    UV Quartz
  • Lightpath:
    5 mm