Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Storage Bins

ESD shelf bin set (Six SOVE3010-4ESD bins), Black, Pack of 6, 3.62×11.81×3.23"
  76015-070
 :   Treston
 :  SBS6-3010-4ESD
Storage Bins
  76015-070
 :  Treston
Storage Bins
Storage Bins

ESD shelf bin set (Six SOVE3010-4ESD bins), Black, Pack of 6, 3.62×11.81×3.23"

 
$180.46
   
Pack of 1
 
 

These high quality, durable Treston bins have been designed for daily use in stores, production facilities and warehouses.

  • Straight sides ensure an efficient use of space
  • Multiple color options make finding the right item easy
  • Open front provides easy access
  • Corrugated base helps retrieve contents, especially smaller sized and flat objects

 : Labels and dividers sold separately, (translucent plastic), labels and blanking piece FP-6M to produce flat surface for adhesive barcode labels on inside rear wall of bins. Contact Avantor representative for more information. Installation is required. Assembly videos available.

 
  • Color:
    Black
  • Description:
    ESD shelf bin set (Six SOVE3010-4ESD bins)
  • Dimensions:
    3.62×11.81×3.23"
  • Size:
    Pack of 6