COLUMN CAPPED ZYMO-SPIN IIICG MINIPREP
Catalog # 76005-156
Supplier: Zymo Research
COLUMN CAPPED ZYMO-SPIN IIICG MINIPREP
Catalog # 76005-156
Supplier: Zymo Research
Supplier Number: C1006-50-G
Frequently Bought Together
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Nunc-Immuno™ MicroWell™ 96-Well Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Difco™ Tryptic Soy Agar, BD Biosciences
Supplier: BD
VWR® Sterile 70% Isopropanol
Supplier: VWR International
BD BBL™ Mueller Hinton II Broth, BD Biosciences
Supplier: BD
Rapid Response™ Plastic Collection Cup with Temperature Strip, BTNX
Supplier: BTNX INC.
NEB®10-beta Competent <i>E.coli</i> (High Efficiency), New England Biolabs
Supplier: New England Biolabs (NEB)
Chloroform-[D₁] (99.8% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
Formvar Carbon Support Film on Square Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
2,2'-Bipyridyl 99+%
Supplier: Thermo Scientific Chemicals
InLab® Intelligent Sensor Management (ISM®) pH/Conductivity Electrodes, METTLER TOLEDO
Supplier: Mettler Toledo
Trypan blue 0.4% in PBS
Supplier: Cytiva