$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
TABLE MAT ROLL 2-LAYER 50FTX3FTX.12IN
76001-634
:
BOTRON COMPANY INC.
TABLE MAT ROLL 2-LAYER 50FTX3FTX.12IN
76001-634
:
BOTRON COMPANY INC.
:
B31350
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International