Order Entry
Canada
Orders LinkContactUsLinkComponent
Serum Vials, Empty

Serum Vials, Empty

Serum vial, empty, 100 ml, Clear, Yes, 10 per pack
Catalog # CA76006-340
Supplier #:  SV100
Serum Vials, Empty
Catalog # CA76006-340
Serum Vials, Empty
Serum Vials, Empty

Serum vial, empty, 100 ml, Clear, Yes, 10 per pack

Price
$170.00
Log in for availability
Pack of 10 (100ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Hardy Diagnostics offers all the products you need to easily assess the risk of microbial contamination of your CSPs (Compounded sterile preparations) according to USP Chapter <797>.

  • Surface sampling
  • Air sampling
  • Sterility tests
  • Quality culture media tested for performance, pH, and sterility
  • Kits are designed to help you follow USP <797> guidelines
  • Kits supply what is needed to test the proficiency and aseptic technique of technicians or pharmacists in a self-contained ready-to-use format
  • Kits are available for low, medium, and high complexity procedures

Empty glass serum vial, terminally autoclaved.

Certifications: Easily assess the risk of microbial contamination of your CSPs (Compounded Sterile Preparations) according to USP Chapter 797.

Specifications
  • Cap included:
    Yes
  • Capacity:
    100 ml
  • Color:
    Clear
  • Description:
    Serum vial, empty
  • Packaging:
    10 per pack
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.