Orders LinkContactUsLinkComponent
Headspace Caps, MicroSolv
Headspace Caps, MicroSolv
Magnetic Screw Cap, 18 mm Precision Threads, 8 mm Hole, Aluminum, Thick Outer Ring/Thin Center for SPME, Silver
Catalog # 75997-582
Supplier #:  95020-26-S
Headspace Caps, MicroSolv
Catalog # 75997-582
Headspace Caps, MicroSolv
Headspace Caps, MicroSolv
Magnetic Screw Cap, 18 mm Precision Threads, 8 mm Hole, Aluminum, Thick Outer Ring/Thin Center for SPME, Silver
Price
$146.72
Log in for availability
Pack of 100
Product Details & Documents
MicroSolv™ headspace caps are available as magnetic and with 4 different septa types to meet needs.
The screw cap eliminates the need for heavy crimping tools and assures that the vials are sealed perfectly every time. The design saves time and money, and ensures a good head space analysis.
Specifications
  • Septum:
    Thick Outer Ring/Thin Center for SPME
  • Color:
    Silver
  • Description:
    Magnetic Screw Cap
  • Material:
    Aluminum
  • Size:
    18 mm Precision Threads, 8 mm Hole