Orders LinkContactUsLinkComponent
Headspace Caps, MicroSolv
Headspace Caps, MicroSolv
Crimp Cap, 20 mm, Aluminum bimetal, Silicone/PTFE, 1.5 mm for SPME Needles (3 mm Outer Ring for Standard HS Vials), Blue
Catalog # 75997-578
Supplier #:  95025-26-4B
Headspace Caps, MicroSolv
Catalog # 75997-578
Headspace Caps, MicroSolv
Headspace Caps, MicroSolv
Crimp Cap, 20 mm, Aluminum bimetal, Silicone/PTFE, 1.5 mm for SPME Needles (3 mm Outer Ring for Standard HS Vials), Blue
Price
$179.39
Log in for availability
Pack of 100
Product Details & Documents
MicroSolv™ headspace caps are available as magnetic and with 4 different septa types to meet needs.
The screw cap eliminates the need for heavy crimping tools and assures that the vials are sealed perfectly every time. The design saves time and money, and ensures a good head space analysis.
Specifications
  • Septum:
    Silicone/PTFE, 1.5 mm for SPME Needles (3 mm Outer Ring for Standard HS Vials)
  • Color:
    Blue
  • Description:
    Crimp Cap
  • Material:
    Aluminum bimetal
  • Size:
    20 mm