Midi Plus™ Pipetting Controller, Sartorius
Midi Plus™ Pipetting Controller
Catalog # CA75840-128
Supplier: Sartorius
: 710931
Jump to:
Product Details & Documents
The lightweight, cordless Midi Plus pipetting controller enables users to aspirate from bottles and tubes without holding their arms and hands in an uncomfortable raised position.
- Precise finger tip control of variable speed ranges
- Hydrophobic autoclavable filter prevents over aspiration
- Low-battery warning
Users can achieve fine-tune speed by applying varying finger pressure to the operating buttons.
The pipetting controller fits all commonly used 1 to 100 ml glass or plastic pipettes.
Specifications
- Description:Midi Plus™ Pipetting Controller
Accessories
Product Information
- Catalog #AvailabilityPrice
- Adapter Set for Midi Plus™, Standard F/1 to 100 mL
Product Information
- Catalog #AvailabilityPrice
- Adapter Set for Midi Plus™, Plus F/10 mL



