Orders LinkContactUsLinkComponent
Millesi Micro Scissors, Sklar
Millesi Micro Scissors, Sklar
159 mm (6¼"), Stainless steel, Curved
Catalog # 75996-208
Supplier:   Sklar
Supplier #:  98-4151
Millesi Micro Scissors, Sklar
Catalog # 75996-208
Supplier:  Sklar
Millesi Micro Scissors, Sklar
Millesi Micro Scissors, Sklar
159 mm (6¼"), Stainless steel, Curved
Price
$2,039.97
New Customer Promotional Price
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sklar's® Millesi micro scissors are used in neurosurgical, ophthalmic and general surgery procedures.
  • Reusable and nonsterile
  • Micro size
The tips of the Millesi micro scissors are used to spread and probe the area of an incision. They are available curved or straight.
Specifications
  • Length:
    159 mm (6¼")
  • Material:
    Stainless steel
  • Curvature:
    Curved
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.