Objectives for REVOLVE Microscopes, Echo
Fluorite objective, 17 mm, 0.13 NA
Catalog # 75992-488
Supplier #: JMAG0042
Jump to:
Discontinued
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:4×
- Description:Fluorite objective
- Aperture:0.13 NA
- Working distance:17 mm