Objectives for REVOLVE Microscopes, Echo
Achromat objective, 5.8 mm, 0.06 NA
Catalog # 75992-484
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0021
Jump to:
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:2×
- Description:Achromat objective
- Aperture:0.06 NA
- Working distance:5.8 mm