Objectives for REVOLVE Microscopes, Echo
Achromat oil objective, 0.15 mm, 1.25 NA
Catalog # 75990-682
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG1005
Jump to:
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:100×
- Description:Achromat oil objective
- Aperture:1.25 NA
- Working distance:0.15 mm