Objectives for REVOLVE Microscopes, Echo
Achromat ELWD phase objective, 3.2 mm, 0.40 NA
75990-654
Supplier #: JMAG0209
Documents
Echo Revolve
Product Literature
- Magnification:20×
- Description:Achromat ELWD phase objective
- Aperture:0.40 NA
- Working distance:3.2 mm