Orders LinkContactUsLinkComponent
Objectives for REVOLVE Microscopes, Echo
Objectives for REVOLVE Microscopes, Echo
Achromat phase objective, 10 mm, 0.25 NA
Catalog # 75990-634
Supplier:   DISCOVER ECHO INC.
Supplier #:  JMAG0105
Objectives for REVOLVE Microscopes, Echo
Catalog # 75990-634
Supplier:  DISCOVER ECHO INC.
Objectives for REVOLVE Microscopes, Echo
Achromat phase objective, 10 mm, 0.25 NA
Price
Inquire For Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Documents
Specifications
  • Magnification:
    10×
  • Description:
    Achromat phase objective
  • Aperture:
    0.25 NA
  • Working distance:
    10 mm