Objectives for REVOLVE Microscopes, Echo
Achromat objective, 10.6 mm, 0.25 NA
Catalog # 75990-632
Supplier: DISCOVER ECHO INC.
: JMAG0104
Jump to:
Product Details & Documents
Specifications
- Magnification:10×
- Description:Achromat objective
- Aperture:0.25 NA
- Working distance:10.6 mm