Scissors, Trimmer, Pattern Cutting, Excelta Corp®
Scissors, trimmer, 2 star quality, 8" (200 mm), Stainless steel AISI 440, 100 mm (4"), Straight
75880-592
: Excelta
: 341
These two star trimmer scissors are used for pattern cutting and general purpose use.
- Ideal scissors for pattern cutting
- Three different sizes to choose from
They are chrome-plated and come in various lengths.
- Description:Scissors, trimmer, 2 star quality
- Length:8" (200 mm)
- Material:Stainless steel AISI 440
- Curvature:Straight
- Blade length:100 mm (4")
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



