Orders LinkContactUsLinkComponent
Animal Identification: Catch a Predator
Animal Identification: Catch a Predator
Catch a Predator Kit II, 6, 24
Catalog # 470225-424
Supplier #:  7006
Animal Identification: Catch a Predator
Catalog # 470225-424
Animal Identification: Catch a Predator
Animal Identification: Catch a Predator
Catch a Predator Kit II, 6, 24
Price
$298.45
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Your students are asked to identify the unknown predator that is killing calves on a nearby ranch.
  • Easy to follow protocols with setup and step-by-step guidance
  • Procedures contain questions allowing better assessment for understanding
  • Allows real life application of common scientific techniques
  • No animal or human body fluids are handled in these experiments
A rancher found hair left behind by a suspect predator. Your studetns are asked to identify the animal responsible by comparing the extracted DNA to that of three known standard samples.
Kit I: Allows students to perform a restriction digest of four DNA samples and compare their sizes via Agarose Gel Electrophoresis.
Kit II: Allows students to amplify four DNA samples via Polymerase Chane Reaction (PCR) and observe visible bands via Agarose Gel Electrophoresis. Master mix (including primers and enzyme) and DNA ladder included.
The kit includes four samples of DNA (plasmids). One representing the DNA of an unknown predator, and three representing known standards for comparison.
Ordering information: Equipment needed but not included: Heat block/water bath for restriction. Thermal cycler for PCR, microcentrifuge; vortex for sample preparation. Electrophoresis equipment/reagents including transilluminator/light box for analysis. Kits come with directions to download instructional material and procedures.
Delivery information: Kits include perishable materials, please provide a delivery date at check-out.
Packaging: Kits contain reagents only.
Specifications
  • Description:
    Catch a Predator Kit II
  • No. of lab groups:
    6
  • No. of students:
    24
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.