Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
2D TANKLINER FIT CNTNR W/DM-33X51CM 38 L
Catalog # 75874-386
Supplier:
AVANTOR FLUID HANDLING, LLC
Expand image
2D TANKLINER FIT CNTNR W/DM-33X51CM 38 L
Catalog # 75874-386
Supplier:
AVANTOR FLUID HANDLING, LLC
Supplier Number:
1003002
Related Information
Documentation
75874-386_20170614-2
75874-386_20170614-2
75874-386_20170614-2
Product Literature
Production insight Solutions for Critical Environments and Manufacturing Processes
VWR Bioprocess Sampling Case Study
VWR Production insight V1
VWR™ Focus Production
Documentation
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning