Freeze Substitution Kit, Electron Microscopy Sciences
Product Details & Documents
For dehydration and fixation of rapidly frozen cells.
- Excellent freeze substitution results in as little as 90 minutes
- Extend the time for larger samples to 3 hours by putting the lid on
Freeze Substitution is a process for low temperature dehydration and fixation of rapidly frozen cells that usually takes days to complete.
With this unique kit researchers are now able to achieve excellent freeze substitution results in as little as 90 minutes for cells of small volume such as bacteria and tissue culture cells. For those cells of greater volume or that have significant diffusion barriers such as cuticles or thick cell walls, one can extend the time to 3 hours simply by putting a lid on the box.
Kit features platform shaker, ice chest, heater block, temperature probe, cryo tubes, and data logger.
Specifications
- Description:Freeze substitution kit with shaker
- Cat. No.:103307-138
- Model:34500
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.