Order Entry
Canada
Orders LinkContactUsLinkComponent
Antibody Diluent, Electron Microscopy Sciences

Antibody Diluent, Electron Microscopy Sciences

Antibody Diluent, 500 ml, Bottle
Catalog # 103307-090
Supplier #:  62715
Antibody Diluent, Electron Microscopy Sciences
Catalog # 103307-090
CAS Number:  
Antibody Diluent, Electron Microscopy Sciences

Antibody Diluent, 500 ml, Bottle

CAS Number:  
Price
$3,074.14
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

These can be used as buffer for diluting primary and secondary antibodies.

  • Reduces non specific binding of the antibodies
  • Minimizes negative effects of disturbing substances
  • Minimizes low or medium affinity cross-reactivities of the antibodies
  • More reliable results

These can be used for diluting antibodies if they were stored even at –20 °C in glycerol, or in refrigerator

For 'trouble-giving' antibodies and in situ PCR applications, this dilutent may be used as a washing buffer, preventing secondary binding of analystes during washing.

When used in pathology, it also greatly reduces non-specific reactivity of human serum components and immunoglobulins in tissue, vessels and cells with mouse antibodies used on section. Excellent for IHC, flow cytometry on fixed cells, western blot and other immune assays.

Specifications

Specifications

  • Pack type:
    Bottle
  • Description:
    Antibody Diluent
  • Size:
    500 ml
  • Cat. No.:
    103307-090
  • Storage temperature:
    4 - 8 °C

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.