Orders LinkContactUsLinkComponent
SmartDoc™ Gel Imaging System, Electron Microscopy Sciences
Gel Imaging System with Blue Light Illumination, 230V
  103306-334
 :  87057-03
SmartDoc™ Gel Imaging System, Electron Microscopy Sciences
  103306-334
SmartDoc™ Gel Imaging System, Electron Microscopy Sciences
SmartDoc™ Gel Imaging System, Electron Microscopy Sciences
Gel Imaging System with Blue Light Illumination, 230V
$2,086.68
Each
Use your smart phone or tablet for high quality imaging.
  • Compatible with all smart phones: iPhone™, Samsung™, LG™, HTC™ and more
  • Instantly share images (text, email or cloud)
  • Adjustable focal distance (for optimum focus)
  • Works with UV and blue light transilluminators
  • Filter for different stain and light sources
  • Blocks ambient light for quality gel images
The SmartDoc™ system is ideal for quickly obtaining quality images of your gels for your records or anaylsis
To use the SmartDoc™, simply place the enclosure over your gel and transilluminator. Lay your smart phone or tablet onto the non-slip, rubber pad, and align the camera lens over the filter. Raising and lowering the platform allows optimization of focus and image size. Filters are available for compatibility with multiple light sources and dyes.
The Blue Light model includes high intensity blue LEDs to excite green dyes. This eliminates the need for hazardous and DNA-damaging UV light.
The SmartDoc™ imaging enclosure includes an orange photo filter for photography of the gels with a smart phone. An extending adapter can be inserted for full dimension imaging of gels up to 15 x 15 cm or removed for close up pictures. The enclosure can also be used directly on any other existing UV or blue light transilluminator. Additional filters are available for imaging gels on a UV transilluminator.
The illumination base emits an intense blue light with a peak output of 460 nm for optimum excitation of nucleic acids stained with green fluorescent dyes (SYBR® Green, SmartGlow™, GelStar™, GelGreen™, etc). The orange cover fits on top of the illumination base and acts as a filter to absorb blue light, allowing visualization of the fluorescing samples. The cover can also be set at an angle for gel access and cutting out DNA bands for further study.
Images can be previewed on the device's display before capturing the picture. Images are easily shared, printed or transferred via text message, email or cloud sharing apps.
Most gel imaging systems cost thousands of dollars as they include a high resolution camera, color display, complicated control panel, computer and more. The SmartDoc™ uses the camera and computer that we already have... our smart phone.
 : UL and CE
  • Description:
    Gel Imaging System with Blue Light Illumination, 230V
Accessories
  •  
    Description
  • 103302-642
    UV Blocking Mat
    Each - $134.24
  •  
    Description
  • 103305-068
    535nm Band Pass Filter for Green Stains
    Each - $375.91
  •  
    Description
  • 103305-070
    590nm Band Pass Filter for EtBr
    Each - $381.33
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.