Orders LinkContactUsLinkComponent
AirChek Badges, Electron Microscopy Sciences
AirChek Badges, Electron Microscopy Sciences
Air Monitoring Badge, Formaldehyde vapor
Catalog # 103306-268
Marketsource
Supplier #:  64472-10
AirChek Badges, Electron Microscopy Sciences
Catalog # 103306-268
Supplier:  
Marketsource
AirChek Badges, Electron Microscopy Sciences
AirChek Badges, Electron Microscopy Sciences
Air Monitoring Badge, Formaldehyde vapor
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BADGE FORMALDEHYDE AIRCHK 8 HRS SMPG PK5 Catalog #78082-102
Product 78082-102 is similar to 103306-268 but may have a different manufacturer and/or packaging.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Monitor vapor levels, simply and effectively.
  • Clips onto technicians clothing
  • Record of name, location, time
Airchek Badges provide an innovative, simple and effective system for monitoring vapor levels, including Formaldehyde, Glutaraldehyde, Xylene, Toluene, and Isopropanol. The badges provide continuous sampling for personal monitoring and allowing the measurements required under Government regulations. One badge provides continuous sampling over 8 hours. A second badge can be used to supply information for analysis of 15-minute peak exposure periods.
Specifications
  • Description:
    Air Monitoring Badge
  • Detectable gas:
    Formaldehyde vapor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)