Saws, Electron Microscopy Sciences
Product Details & Documents
Three styles of manual bone saws are available.
- Broad bladed Satterlee Saw
- Thinner rounded top Charriere Saw
- Solid handle Langenbech Metacarpal Saw
The broad bladed Satterlee Saw (A), the thinner rounded tip Charriere Saw (B), and the solid handle Langenbech Metacarpal Saw (C). Each saw has serrated teeth.
Specifications
- Description:Charriere Saw, 267 mm Long
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.