Orders LinkContactUsLinkComponent
Whipple Grid Graticules, Electron Microscopy Sciences
Whipple Grid Graticules, Electron Microscopy Sciences
NE29 Graticule, 27.0 mm
Catalog # 103305-438
Supplier #:  68009-27
Whipple Grid Graticules, Electron Microscopy Sciences
Catalog # 103305-438
Whipple Grid Graticules, Electron Microscopy Sciences
Whipple Grid Graticules, Electron Microscopy Sciences
NE29 Graticule, 27.0 mm
Price
$415.08
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Whipple grid graticules are used for drinking water particle analysis and for other particle counting.
  • Whipple grid 100 squares in 7 mm area
  • Surface chrome image
Grid showed ratio of full square to smallest is 50:1 and area is 2500:1.
Specifications
  • Description:
    NE29 Graticule
  • Size:
    27.0 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)