Order Entry
Canada
Orders LinkContactUsLinkComponent
EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences

EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences

EDTA Buffer, 100 mL, 8.5
Catalog # 103302-792
Supplier #:  64142-09
EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences
Catalog # 103302-792
CAS Number:  
EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences
EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences

EDTA Buffer, 100 mL, 8.5

CAS Number:  
Price
$191.72
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This buffer is intended for heat-induced antigen retriever on formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies.

In IHC most commonly used fixative like formalin mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC

Preparation of Working Solution: Dilute this 10X buffer as needed (90 mL of deionized or distilled water+10 mL of this buffer) mix well, 1X buffer can be stored at 2 to 8 °C.

The use of this buffer on FFPE tissue section improves accessibility of antibodies to tissue antigens.

Specifications

Specifications

  • Description:
    EDTA Buffer
  • Size:
    100 mL
  • pH:
    8.5

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.