Chemical Containers, Electron Microscopy Sciences
Product Details & Documents
Relied on to transport dust samples from the moon, these injection-molded polyethylene containers are ideal for small quantities of liquids or solids.
- Made of injection-molded polyethylene
- Screw caps feature a leakproof inner seal and prevent leakage and spill
- Easy to clean
Specifications
- Ext.Ø:32 mm
- Capacity:30 ml
- Description:Container
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)