Orders LinkContactUsLinkComponent
Athene Hexagonal Grids, Electron Microscopy Sciences
Athene Hexagonal Grids, Electron Microscopy Sciences
Hexagonal, Nickel, 100 Mesh
Catalog # 103302-684
Supplier #:  AH100-NI
Athene Hexagonal Grids, Electron Microscopy Sciences
Catalog # 103302-684
Athene Hexagonal Grids, Electron Microscopy Sciences
Athene Hexagonal Grids, Electron Microscopy Sciences
Hexagonal, Nickel, 100 Mesh
Price
166,16 $
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These hexagonal athene grids are used in transmission electron microscope.
  • Copper and nickel grids
  • 100, and 400 mesh grids
  • Good handling characteristics
  • Individually inspected before being packed
3.05 mm diameter.
Specifications
  • Description:
    Hexagonal
  • Material:
    Nickel
  • Mesh:
    100 Mesh
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)