Athene Hexagonal Grids, Electron Microscopy Sciences
These hexagonal athene grids are used in transmission electron microscope.
- Copper and nickel grids
- 100, and 400 mesh grids
- Good handling characteristics
- Individually inspected before being packed
3.05 mm diameter.
- Description:Hexagonal
- Material:Copper
- Mesh:400 Mesh
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)