Scratch Brushes with Metal Ferrules, Electron Microscopy Sciences
Product Details & Documents
These scratch brushes with pen shaped handles are used for fine deburring, polishing, as well as rust removal and the cleaning and removal of deposited wax.
- Available with steel, nylon, fiberglass or brass
- With metal ferrules for improved durability
- Overall length is 41/4" and the brush is 1/8" wide
Packaging: Refills are available and are packaged 12/box.
Specifications
- Bristle material:Fiberglass
- Description:Fiberglass Ferrules
- Total length:4 1/4"
- Brush head width:1/8"
Accessories
Product Information
- Catalog #AvailabilityPrice
- Brush Refill, Nylon Ferrules
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)