Orders LinkContactUsLinkComponent
Athene Thick/Thin Bar with Center Mark Grids, Electron Microscopy Sciences
Athene Thick/Thin Bar with Center Mark Grids, Electron Microscopy Sciences
Thick and Thin Bar with Center Mark, Copper, 3.05 mm, 200
Catalog # 103302-116
Marketsource
Supplier #:  ATT200-CU
Athene Thick/Thin Bar with Center Mark Grids, Electron Microscopy Sciences
Catalog # 103302-116
Marketsource
Athene Thick/Thin Bar with Center Mark Grids, Electron Microscopy Sciences
Athene Thick/Thin Bar with Center Mark Grids, Electron Microscopy Sciences
Thick and Thin Bar with Center Mark, Copper, 3.05 mm, 200
Price
$155.82
Log in for availability
Each Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Athene grids are renowned for the highest quality mesh for transmission electron microscope.
  • Good handling characteristics
  • Individually inspected before being packed
Specifications
  • Description:
    Thick and Thin Bar with Center Mark
  • Dimensions:
    3.05 mm
  • Material:
    Copper
  • Mesh:
    200
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)