Stainless Steel Baskets with Multiple Compartments, Electron Microscopy Sciences
Product Details & Documents
These stainless steel baskets with lids and multiple compartments are used for critical point drying.
- Single or multiple compartments
These baskets are either 15 or 23 mm in diameter.
Specifications
- Description:4 Compartment Tissue Basket with Lid
- I.D.×H:25 mm×23 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)