Order Entry
Canada
Orders LinkContactUsLinkComponent
Membrane Boxes, Electron Microscopy Sciences

Membrane Boxes, Electron Microscopy Sciences

Membrane box, square, 200×200×50 mm, 154×154×40 mm
Catalog # 103302-034
Marketsource
Supplier #:  64304-26
Membrane Boxes, Electron Microscopy Sciences
Catalog # 103302-034
Marketsource
Membrane Boxes, Electron Microscopy Sciences
Membrane Boxes, Electron Microscopy Sciences

Membrane box, square, 200×200×50 mm, 154×154×40 mm

Price
$131.12
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These unique membrane boxes are for the transfer, storage and shipping of many delicate items. Once your sensitive item is placed in the box the elastic membrane conforms around the enclosed object and protects it from any movement.

  • Protects items from any movement
  • Double elastic membrane on the top and bottom offers security
  • Available in round, rectangular and square shapes
Specifications
  • Description:
    Membrane box, square
  • Dimensions:
    200×200×50 mm
  • Interior dimensions:
    154×154×40 mm

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.