Orders LinkContactUsLinkComponent
Chemical Containers, Electron Microscopy Sciences
Chemical Containers, Electron Microscopy Sciences
120 ml, Container, 52 mm
Catalog # 103302-004
Supplier #:  64299-44
Chemical Containers, Electron Microscopy Sciences
Catalog # 103302-004
Chemical Containers, Electron Microscopy Sciences
Chemical Containers, Electron Microscopy Sciences
120 ml, Container, 52 mm
Price
$239.67
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Relied on to transport dust samples from the moon, these injection-molded polyethylene containers are ideal for small quantities of liquids or solids.
  • Made of injection-molded polyethylene
  • Screw caps feature a leakproof inner seal and prevent leakage and spill
  • Easy to clean
Specifications
  • Ext.Ø:
    52 mm
  • Capacity:
    120 ml
  • Description:
    Container
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.