VWR® Certified Reference Materials, Oxide
2 kg, Gold, 20.00 - 50.00 ppm, Au, Oxide, Jar
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
Supplier #: 21001111
VWR® Certified Reference Materials, Oxide
Catalog # CA75852-356
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
CAS Number:
Jump to:
Product Details & Documents
KLEN's CRMs are certified products containing a range of analytes including Au, Ag, Pt, and Pd at various ppm levels
Ordering information: Other sizes (30g, 50g and 150g sachets) are available, please contact your Avantor sales representative for more information.
Specifications
Specifications
- Concentration:20.00 - 50.00 ppm
- Matrix:Oxide
- Pack type:Jar
- Size:2 kg
- Analyte:Au
- Element:Gold
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



