VWR® Certified Reference Materials, Oxide
2 kg, Gold, 10.00 - 20.00 ppm, Au, Oxide, Jar
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
Supplier #: 21001101
VWR® Certified Reference Materials, Oxide
Catalog # CA75852-354
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
CAS Number:
Jump to:
Product Details & Documents
KLEN's CRMs are certified products containing a range of analytes including Au, Ag, Pt, and Pd at various ppm levels
Ordering information: Other sizes (30g, 50g and 150g sachets) are available, please contact your Avantor sales representative for more information.
Specifications
Specifications
- Concentration:10.00 - 20.00 ppm
- Matrix:Oxide
- Pack type:Jar
- Size:2 kg
- Analyte:Au
- Element:Gold



