Orders LinkContactUsLinkComponent
Dissolution Cannula Filters, Quality Lab Accessories | Avantor
Dissolution Cannula Filters, Quality Lab Accessories
20 Micron Filter Discs, UHMW Polyethylene, Jar
Catalog # 75848-914
Supplier:   QLA
Fournisseur #:  FIL020-DK
Dissolution Cannula Filters, Quality Lab Accessories
Catalog # 75848-914
Supplier:  QLA
Dissolution Cannula Filters, Quality Lab Accessories
Dissolution Cannula Filters, Quality Lab Accessories
20 Micron Filter Discs, UHMW Polyethylene, Jar
Price
$482.41
Log in for availability
Case of 1,000
Product Details & Documents
Only QLA manufactures pure quality filters which are guaranteed chromium and heavy metal free.
  • Made of the purest and highest quality UHMW Polyethylene (and PVDF where applicable)
  • All filters are made in the USA
  • Available in 100 filter packs or 1000 filter jars
Filters for most sampling methods are available in numerous micron ratings and three main types: cannula filters, filter discs, and filter tips. The PureQuality certification program guarantees filters not to have any chromium, heavy metals, material contaminants, or inorganic elemental interference. In addition, they have been formulated to provide the specified micron rating and verified by porisometer testing. All filters are USP compliant where applicable, and meet machine manufacturer specifications.
Certifications: All filters are supplied with a Certificate of Conformance (COC) stating that they are USP compliant, where applicable, and that they meet machine manufacturer specifications.
Specifications
  • Description:
    20 Micron Filter Discs, UHMW Polyethylene
  • Packaging:
    Jar
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.