Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 $
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ACETIC ACID ISOOCTANE CLASS: 3 60:40 4L
Catalog # RCR0161000-4C1
Supplier:
Ricca Chemical
SDS
Certificates
Expand image
ACETIC ACID ISOOCTANE CLASS: 3 60:40 4L
Catalog # RCR0161000-4C1
Supplier:
Ricca Chemical
Supplier Number:
R0161000-4C1
SDS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
SDS
ghs_R0161000.pdf
Documentation
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International