Glass Stirring Rods
6 mm, 254 mm
Catalog # 470200-955
Supplier: United Scientific Supplies
Supplier #: GSR010
Jump to:
Product Details & Documents
Glass stirring rod features fire polished flint glass.
- Rounded ends
Stirring rods are made of soft flint glass, with both ends fire-polished and rounded.
Specifications
- Ø:6 mm
- Length:254 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



