Biox Freezer Boxes
Freezer Box, Polycarbonate, 100, 5 cm (2")
Catalog # 75799-924
Supplier: TWD Scientific
: BX90-9210-1
Jump to:
Product Details & Documents
Freezer Boxes are designed to provide easy inventory management of cryogenic vials used in tissue culture, pharmaceutical research, and other biotechnology labs.
- Store in mechanical freezers (PC/PP) or liquid nitrogen (PC)
- Vented boxes allow rapid cooling and full drainage
- Chemically resistant to alcohols and mild solvents
- Grid on lid for easy identification
Plastic freezer boxes are specifically designed for Liquid nitrogen (PC) and mechanical freezer (PP) storage.
Specifications
- No. of Holes:100
- Description:Freezer Box, Polycarbonate
- Height:5 cm (2")



