Biox Cell Strainers
100 µm, Green
75799-944
: BX15-1100
Cell Strainers feature a strong nylon mesh for optimal performance in a variety of applications.
- Polypropyelene frame with nylon mesh
- Gamma radiation sterilized
- Color coded for easy pore size identification
- Individually wrapped
- Applications in cell culture and filtration of various cells
Perfect fit for 50 ml centrifuge tubes and color coded according to the pore size.
- Pore size:100 µm
- Color:Green



