Biox Cell Strainers
70 µm, Orange
75799-942
: BX15-1070
Cell Strainers feature a strong nylon mesh for optimal performance in a variety of applications.
- Polypropyelene frame with nylon mesh
- Gamma radiation sterilized
- Color coded for easy pore size identification
- Individually wrapped
- Applications in cell culture and filtration of various cells
Perfect fit for 50 ml centrifuge tubes and color coded according to the pore size.
- Pore size:70 µm
- Color:Orange
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



