Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
AB GT A-HU SECY SPECTRA650 250UL F/FLUOR
Catalog # CA75794-676
Supplier:
Azure Biosystems
SDS
Expand image
AB GT A-HU SECY SPECTRA650 250UL F/FLUOR
Catalog # CA75794-676
Supplier:
Azure Biosystems
Supplier Number:
AC2167
SDS
Related Information
Documentation
Product Literature
Azure Biosystems Western Blotting Workflow
SDS
ghs_AC2167
Documentation
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International