Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 $
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
E. COLI O 78: K 80 ANTISERA 3ML FOR IHC
Catalog # 75785-634
Supplier:
PRO-LAB DIAGNOSTICS INC CA
SDS
Expand image
E. COLI O 78: K 80 ANTISERA 3ML FOR IHC
Catalog # 75785-634
Supplier:
PRO-LAB DIAGNOSTICS INC CA
Supplier Number:
PLA526
SDS
Related Information
Documentation
SDS
ghs_PLA526.pdf
Documentation
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International