Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ANTISERA SHIGELLA BOYDII TYPE 14 3ML IHC
Catalog # 75785-600
Supplier:
PRO-LAB DIAGNOSTICS INC CA
SDS
Expand image
ANTISERA SHIGELLA BOYDII TYPE 14 3ML IHC
Catalog # 75785-600
Supplier:
PRO-LAB DIAGNOSTICS INC CA
Supplier Number:
PLA424
SDS
Related Information
Documentation
SDS
ghs_PLA424.pdf
Documentation
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma