0,00 $
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
SALMONELLA H: Z4 Z23 3ML PHS INVERSION
75785-442
:
PRO-LAB DIAGNOSTICS INC CA
SALMONELLA H: Z4 Z23 3ML PHS INVERSION
75785-442
:
PRO-LAB DIAGNOSTICS INC CA
:
PLA256
ghs_PLA256.pdf
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH