$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
COLUMN HPLC CAPY POLYRP-NP 3 UM 4 X10MM
103056-428
:
Sepax Technologies
COLUMN HPLC CAPY POLYRP-NP 3 UM 4 X10MM
103056-428
:
Sepax Technologies
:
262003-4001
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International