Orders LinkContactUsLinkComponent
Cap Inserts for Cryogenic Vials, Globe Scientific
Cap Inserts for Cryogenic Vials, Globe Scientific
Cap inserts for cryogenic vials, Violet
Catalog # 75784-238
Supplier:   Globe Scientific
Supplier #:  30079V
Cap Inserts for Cryogenic Vials, Globe Scientific
Catalog # 75784-238
Supplier:  Globe Scientific
Cap Inserts for Cryogenic Vials, Globe Scientific
Cap Inserts for Cryogenic Vials, Globe Scientific
Cap inserts for cryogenic vials, Violet
Price
$121.95
New Customer Promotional Price
Log in for availability
Pack of 1,000
Product Details & Documents
Produced from polypropylene, cap inserts are available in assorted colors and are ideal for color coding and sample identification
  • Polypropylene (PP)
  • Designed for use with globe scientific CryoClear cryogenic vials
  • Suitable for use on both internal and extenrally threaded caps
The inserts nest easily into place on both internal and external caps.
Ordering information: Cap inserts are packaged 1000 per bag.
Specifications
  • Color:
    Violet
  • Description:
    Cap inserts for cryogenic vials