Order Entry
Canada
Orders LinkContactUsLinkComponent
Biox Cardboard Freezer Boxes

Biox Cardboard Freezer Boxes

81, 7.6 cm (3")
Catalog # 75780-330
Supplier:   TWD Scientific
Supplier #:  BX90-2381
Biox Cardboard Freezer Boxes
Catalog # 75780-330
Supplier:  TWD Scientific
Biox Cardboard Freezer Boxes
Biox Cardboard Freezer Boxes

81, 7.6 cm (3")

Price
$638.35
Log in for availability
Case of 100
Product Details & Documents

Premium cardboard freezer boxes with a water-resistant coating.

  • Chemically resistant to alcohols and mild organic solvents
  • Temperature range: –196 to 121°C (–320 to 250 °F)
  • Accommodates 0.5 ml, 1.5 ml, or 2.0 ml microtubes
  • Suitable for liquid nitrogen storage

Boxes feature a water-resistant coating for improved durability.

Specifications
  • No. of Holes:
    81
  • Height:
    7.6 cm (3")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.